Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Bromo-2-chloronicotinate 98.0+%, TCI America™
SDP

Catalog No. M25135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25135G Supplier TCI America Supplier No. M25135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity building block for chemical synthesis with a minimum purity of 98.0% by GC.

Chemical Identifiers

CAS 78686-79-0
Molecular Formula C7H5BrClNO2
Molecular Weight (g/mol) 250.476
MDL Number MFCD03844848
InChI Key MOMQDEDQGJAKII-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-chloronicotinate, methyl5-bromo-2-chloronicotinate, 5-bromo-2-chloro-nicotinic acid methyl ester, methyl 5-bromo-2-chloro-3-pyridinecarboxylate, 5-bromo-2-chloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-2-chloro-, methyl ester, methyl 5-bromo-2-chloro-pyridine-3-carboxylate, 5-bromo-2-chloro-3-methoxycarbonyl pyridine, pubchem12966, acmc-209pf0
PubChem CID 2763343
IUPAC Name methyl 5-bromo-2-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1Cl)Br

Specifications

Melting Point 52°C
Quantity 5 g
Formula Weight 250.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Bromo-2-chloronicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.