Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-bromo-2-(methylsulfanyl)-4-pyrimidinecarboxylate, 97%, Thermo Scientific™

Catalog No. mo01274da
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Catalog No. MO01274DA

Supplier: Thermo Scientific Chemicals MO01274DA

Only null left
Add to Cart

Chemical Identifiers

CAS 50593-91-4
Molecular Formula C7H7BrN2O2S
Molecular Weight (g/mol) 263.11
MDL Number MFCD05664428
InChI Key MYZJIEWTRJTWCD-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester
PubChem CID 2794729
SMILES COC(=O)C1=NC(SC)=NC=C1Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate
Formula Weight 263.11
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-2-(methylsulfanyl)-4-pyrimidinecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.