Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Bromonicotinate 98.0+%, TCI America™
SDP

Catalog No. M20081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20081G 1 g
M20085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20081G Supplier TCI America Supplier No. M20081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29681-44-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00173668
InChI Key AAJZXPWBILCHAW-UHFFFAOYSA-N
Synonym methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
PubChem CID 699336
IUPAC Name methyl 5-bromopyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1)Br

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 1 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Bromonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.