Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-(chloromethyl)-2-furoate, 95%, Thermo Scientific™

Catalog No. BLT00257DA
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BLT00257DA 1 g
BLT00257EA 10 g
BLT00257EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BLT00257DA Supplier Thermo Scientific Chemicals Supplier No. BLT00257DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2144-37-8
Molecular Formula C7H7ClO3
Molecular Weight (g/mol) 174.58
MDL Number MFCD00020922
InChI Key PWXMEBZOKUPCST-UHFFFAOYSA-N
Synonym methyl 5-chloromethyl-2-furoate, methyl 5-chloromethyl furan-2-carboxylate, methyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-furan-2-carboxylic acid methyl ester, 2-furancarboxylic acid, 5-chloromethyl-, methyl ester, 5-chloromethylfuran-2-carboxylic acid methyl ester, pubchem24261, enamine_005466, acmc-1ck5d
PubChem CID 75065
IUPAC Name methyl 5-(chloromethyl)furan-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)CCl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 174.58
Percent Purity 95%
Chemical Name or Material Methyl 5-(chloromethyl)-2-furoate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.