Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Hexenoate 98.0+%, TCI America™
SDP

Catalog No. H087225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H087225G 25 g
H08725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H087225G Supplier TCI America Supplier No. H087225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2396-80-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00671538
InChI Key ASKDFGVMJZMYEM-UHFFFAOYSA-N
Synonym 5-Hexenoic Acid Methyl Ester
PubChem CID 520082
IUPAC Name methyl hex-5-enoate
SMILES COC(=O)CCCC=C

Specifications

Color Colorless
Boiling Point 157°C
Quantity 25 g
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Hexenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.