Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Hydroxyanthranilate 98.0+%, TCI America™
SDP

Catalog No. M27961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27961G 1 g
M27965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27961G Supplier TCI America Supplier No. M27961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1882-72-0
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD05863263
InChI Key DWBKSTKVIIRFHL-UHFFFAOYSA-N
Synonym 5-Hydroxyanthranilic Acid Methyl Ester, Methyl 2-Amino-5-hydroxybenzoate, 2-Amino-5-hydroxybenzoic Acid Methyl Ester
PubChem CID 819940
IUPAC Name methyl 2-amino-5-hydroxybenzoate
SMILES COC(=O)C1=C(N)C=CC(O)=C1

Specifications

Melting Point 160°C
Quantity 1 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Hydroxyanthranilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.