Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Iodosalicylate 98.0+%, TCI America™
SDP

Catalog No. I053925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I053925G 25 g
I05395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I053925G Supplier TCI America Supplier No. I053925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4068-75-1
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00016462
InChI Key NRSWJTRJHPRZMH-UHFFFAOYSA-N
Synonym methyl 5-iodosalicylate, 5-iodosalicylic acid methyl ester, 2-hydroxy-5-iodobenzoic acid methyl ester, benzoic acid, 2-hydroxy-5-iodo-, methyl ester, methyl 2-hydroxy-5-iodobenzenecarboxylate, salicylic acid, 5-iodo-, methyl ester, methyl5-iodosalicylate, methyl-5-iodosalicylate, acmc-209jg0, methyl2-hydroxy-5-iodobenzoate
PubChem CID 248910
IUPAC Name methyl 2-hydroxy-5-iodobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)I)O

Specifications

Melting Point 77°C
Color White
Quantity 25 g
Formula Weight 278.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Iodosalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.