Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Methoxysalicylate 98.0+%, TCI America™
SDP

Catalog No. M21275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21275G 5 g
M212725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21275G Supplier TCI America Supplier No. M21275G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2905-82-0
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00017185
InChI Key DFNBGZODMHWKKK-UHFFFAOYSA-N
Synonym 5-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-5-methoxybenzoate, 2-Hydroxy-5-methoxybenzoic Acid Methyl Ester
PubChem CID 4072341
IUPAC Name methyl 2-hydroxy-5-methoxybenzoate
SMILES COC(=O)C1=C(O)C=CC(OC)=C1

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Methoxysalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.