Learn More
AA BLOCKS LLC Methyl 6-chloro-1h-indole-5-carboxylate, 5g
Supplier:  AA BLOCKS LLC AA001SO15G
Encompass_Preferred
Methyl 6-chloro-1h-indole-5-carboxylate | CAS Number:162100-83-6 | Molecular Formula: C10H8ClNO2 | Molecular Weight: 209.6290 | MDL Number: MFCD11976069 | SMILES: COC(=O)c1cc2cc[nH]c2cc1Cl
Catalog No. NC2682942