Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-Chloronicotinate 98.0+%, TCI America™
SDP

Catalog No. M198025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M198025G 25 g
M19805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M198025G Supplier TCI America Supplier No. M198025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73781-91-6
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00023420
InChI Key RMEDXVIWDFLGES-UHFFFAOYSA-N
Synonym methyl 6-chloronicotinate, 6-chloronicotinic acid methyl ester, methyl6-chloronicotinate, methyl 2-chloropyridine-5-carboxylate, 3-pyridinecarboxylic acid, 6-chloro-, methyl ester, methyl 6-chloro-3-pyridinecarboxylate, 6-chloro-3-pyridinecarboxylic acid methyl ester, pubchem13584, methyl-6-chloronicotinate, methyl 6-chloro-nicotinate
PubChem CID 254249
IUPAC Name methyl 6-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)Cl

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 25 g
Formula Weight 171.58
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Chloronicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.