Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-fluoronicotinate, 95%, Thermo Scientific Chemicals

Catalog No. AAH61302MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61302MD 250 mg
AAH6130203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61302MD Supplier Thermo Scientific Chemicals Supplier No. H61302MD

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1427-06-1
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD03095098
InChI Key HLYBWNNPVXFCPZ-UHFFFAOYSA-N
PubChem CID 13758765
IUPAC Name methyl 6-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)F

Specifications

Melting Point 48°C to 52°C
Flash Point 99°C (210°F)
Quantity 250 mg
Synonym methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate
Formula Weight 155.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 6-fluoronicotinate
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.