Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-(Hydroxymethyl)nicotinate 97.0+%, TCI America™
SDP

Catalog No. M27501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27501G 1 g
M2750200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27501G Supplier TCI America Supplier No. M27501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56026-36-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00643821
InChI Key QVLONHSXRWCCAE-UHFFFAOYSA-N
Synonym methyl 6-hydroxymethyl nicotinate, 6-hydroxymethyl-nicotinic acid methyl ester, methyl 6-hydroxymethyl pyridine-3-carboxylate, 6-hydroxymethylnicotinic acid methyl ester, 6-hydroxymethyl-nicotinicacidmethylester, methyl 6-hydroxymethylnicotinate, 3-pyridinecarboxylic acid, 6-hydroxymethyl-, methyl ester, 3-pyridinecarboxylicacid, 6-hydroxymethyl-, methyl ester, pubchem19720, methylhydroxymethylnicotinate
PubChem CID 789414
IUPAC Name methyl 6-(hydroxymethyl)pyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)CO

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 1 g
Formula Weight 167.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-(Hydroxymethyl)nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.