Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-Quinolinecarboxylate 97.0+%, TCI America™
SDP

Catalog No. M168725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M168725G 25 g
M16875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M168725G Supplier TCI America Supplier No. M168725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38896-30-9
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00956766
InChI Key XSRWQTDEIOHXSL-UHFFFAOYSA-N
Synonym methyl 6-quinolinecarboxylate, 6-quinolinecarboxylic acid, methyl ester, methylquinoline-6-carboxylate, 6-quinolinecarboxylic acid methyl ester, methyl 6-quinoline carboxylate, quinoline-6-carboxylic acid methyl ester, pubchem19578, maybridge1_005415, acmc-209j2g, ksc536k0t
PubChem CID 736812
IUPAC Name methyl quinoline-6-carboxylate
SMILES COC(=O)C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point 85°C
Quantity 25 g
Formula Weight 187.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Quinolinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.