Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™

Catalog No. CC39322DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC39322DA 1 g Amber Glass Bottle
CC39322EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC39322DA Supplier Thermo Scientific Chemicals Supplier No. CC39322DA
Add to Cart

Chemical Identifiers

CAS 118465-49-9
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD07772830
InChI Key YBIAQTPXONLBJG-UHFFFAOYSA-N
Synonym methyl 6h-thieno 2,3-b pyrrole-5-carboxylate, methyl thiopheno 3,2-d pyrrole-5-carboxylate, 5-methoxycarbonyl-6h-thieno 2,3-b pyrrole
PubChem CID 7164601
IUPAC Name methyl 6H-thieno[2,3-b]pyrrole-5-carboxylate
SMILES COC(=O)C1=CC2=C(N1)SC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 181.21
Percent Purity 97%
Chemical Name or Material Methyl 6H-thieno[2, 3-b]pyrrole-5-carboxylate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.