Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl acetate, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018921 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8921 5 mL
11-102-0230 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8921 Supplier MilliporeSigma Supelco Supplier No. 459975MLF
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 936°F
CAS 79-20-9
Vapor Pressure 165 mmHg (20°C)
Vapor Density 2.55 (vs. air)
Density 0.934 g/mL (at 25°C)
Flash Point -13°C
Physical Form Neat
Linear Formula CH3 COOCH3
MDL Number MFCD00008711
Melting Point -98°C
Boiling Point 57°C to 58°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 74.08
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.