Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Alcohol, Electronic Grade, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18604408 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 x 1 gal.
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-408 4 x 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-408 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M12391GLCS4
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M1239, 67-56-1, CH3OH

Spectrum™ Chemical Methyl Alcohol, Electronic Grade is also known as wood alcohol and methanol. Spectrum's ELECTRONIC GRADE products are suitable for use in electronic component manufacturing. These materials meet or exceed the specifications as listed in Reagent Chemicals published by the American Chemical Society.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Acidity 0.3 μeq/g
CAS Max % 100%
Alkalinity 0.1 μeq/g
Quantity 4 x 1 gal.
Packaging Poly Bottle
Residue after Evaporation 5 ppm
Water 0.05%
UN Number UN1230
Boiling Point 64°C to 65°C
Percent Purity 99.9%
Grade Electronic
Chemical Name or Material Methyl Alcohol
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.