Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18611673 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
200 L
Packaging:
Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-673 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-611-673 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HP706200LTBL

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 1
Appearance To Pass Test
Quantity 200 L
Packaging Drum
Residue after Evaporation 5 ppm
Titratable Acid 0.3 μeq/g
Titratable Base 0.2 μeq/g
Water 0.001
Percent Purity 0.999
Grade HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.