Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Alcohol, NF, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18604413 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-413 500 mL Amber Glass Bottle
18-604-414 6 x 500 mL Amber Glass Bottle
18-604-415 4 L Amber Glass Bottle
18-604-416 4 x 4 L Amber Glass Bottle
18-604-417 20 L Steel Can
18-604-418 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-413 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ME151500MLGL
Add to Cart
Add to Cart

ME151, 67-56-1, CH3OH

Spectrum™ Chemical Methyl Alcohol, NF is as a solvent excipient for pharmaceutical products. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100%
Alkalinity 3 ppm
Quantity 500 mL
Packaging Amber Glass Bottle
UN Number UN1230
Boiling Point 64°C to 65°C
Percent Purity 99.5%
Grade NF
Chemical Name or Material Methyl Alcohol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.