Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Alcohol, Purified, Spectrum™ Chemical
SDP

Catalog No. 18604424 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
4 x 4 L
200 L
Packaging:
Fiber Drum
Glass Bottle
Plastic Bottle
Steel Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-424 200 L Fiber Drum
18-604-419 4 L Glass Bottle
18-604-422 4 L Plastic Bottle
18-604-420 4 x 4 L Glass Bottle
18-604-423 4 x 4 L Plastic Bottle
18-604-421 20 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-424 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1245200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M1245, 67-56-1, CH3OH

Spectrum™ Chemical Methyl Alcohol, Purified is a colorless, flammable substance used in chemical labs as a polar protic solvent in organic synthesis, as well as a carrier (mobile phase) solvent in high pressure liquid chromatography (HPLC). Methanol is the simplest type of alcohol and was, at one point, chiefly made from the distillation of wood, and is commonly referred to as wood alcohol, wood spirits or methyl alcohol.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100%
Quantity 200 L
Packaging Fiber Drum
UN Number UN1230
Boiling Point 64°C to 65°C
Grade Purified
Chemical Name or Material Methyl Alcohol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.