Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl alpha-D-Galactopyranoside Monohydrate 98.0+%, TCI America™
SDP

Catalog No. M104725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M104725G 25 g
M10475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M104725G Supplier TCI America Supplier No. M104725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34004-14-3
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00150748
InChI Key HOVAGTYPODGVJG-PZRMXXKTSA-N
Synonym methyl alpha-d-galactopyranoside monohydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol hydrate, methyl-alpha-d-galactopyranoside, methyl ?-galactoside hydrate, methyl a-d-galactopyranoside,monohydrate, methyl alpha-d-galactopyranoside hydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxyoxane-3,4,5-triol hydrate
PubChem CID 2724703
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 108°C
Color White
Quantity 25 g
Formula Weight 194.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl alpha-D-Galactopyranoside Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.