Learn More
Cayman Chemical Methyl Arachidonyl Fluorophosphonate, 188404-10-6, 5 mg
SDP
Supplier:  Cayman Chemical 706605
Encompass_Preferred
A selective, active-site directed, irreversible inhibitor of cPLA2 and iPLA2; inhibits A23187-induced arachidonic acid release from human platelets (IC50 = 0.6 µM) and iPLA2 release from P388D1 cells (IC50 = 0.5 µM); a potent inhibitor of FAAH (IC50 = 2.5 nM); binds to the CB1 receptor in rat brain membrane preparations(IC50 = 20 nM)
Catalog No. NC0435771
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.