Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl azetidine-3-carboxylate hydrochloride | 100202-39-9 | MFCD01861758 | 99.7% | 151.59 | C5H10ClNO2 | 1 G
SDP

Supplier:  Medchemexpress LLC HY336151G

Encompass_Preferred

Methyl azetidine-3-carboxylate hydrochloride is a methyl-substituted azetidine carboxylate hydrochloride salt supplied as a white to off-white solid for research use. It is used as a non-cleavable ADC linker and as an alkyl chain-based PROTAC linker in medicinal chemistry and conjugation applications.

  • High purity (99.7%) suitable for synthesis and conjugation.
  • Solid form, white to off-white, easy to handle and weigh.
  • Storage: sealed at 4°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Supplied in small research pack sizes (example: 1 G).
  • Chemical identifiers: CAS 100202-39-9; formula C5H10ClNO2; molecular weight 151.59.

Catalog No. 50-002-03204


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.