Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Benzenesulfinate 98.0+%, TCI America™
SDP

Catalog No. M297125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M297125G 25 g
M29715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M297125G Supplier TCI America Supplier No. M297125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 670-98-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD00674051
InChI Key PSNSVDSRLUYDKF-UHFFFAOYSA-N
Synonym methylbenzenesulfinate, benzenesulfinic acid methyl ester, methyl phenylsulfinate, methyl benzene sulfinate, benzenesulfinic acid,methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, methoxy phenylsulfinyl, acmc-20mem2, acmc-20mg8f
PubChem CID 2759784
IUPAC Name methyl benzenesulfinate
SMILES COS(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 63°C
Quantity 25 g
Formula Weight 156.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Benzenesulfinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.