Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Benzoylformate, Spectrum™ Chemical
SDP

Catalog No. 18621057 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-057 25 g
18-621-058 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-621-057 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M159525GM
Add to Cart
Add to Cart

Synonym: Methyl 2-Oxo-2-Phenylacetate, Methyl Phenylglyoxalate, Methyl Oxophenylacetate

Chemical Identifiers

CAS 15206-55-0

Specifications

Boiling Point 251°C
Packaging Amber Bottle
Quantity 25 g
Percent Purity 97%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.