Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl chloroacetate, 98%

Catalog No. AC148630100
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148630100 10 L Plastic drum
AC148630010 1 L Glass bottle
AC148630025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC148630100 Supplier Thermo Scientific Chemicals Supplier No. 148630100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 96-34-4 | C3H5ClO2 | 108.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-34-4
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00000931
InChI Key QABLOFMHHSOFRJ-UHFFFAOYSA-N
Synonym methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
PubChem CID 7295
IUPAC Name methyl 2-chloroacetate
SMILES COC(=O)CCl

Specifications

Melting Point -33.0°C
Color Colorless
Density 1.2300g/mL
Boiling Point 130.0°C to 132.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.4210 to 1.4230
Linear Formula ClCH2CO2CH3
Quantity 10 L
Beilstein 02,197
Merck Index 15,6113
Specific Gravity 1.23
Solubility Information Solubility in water: 28g/L (20°C). Other solubilities: soluble in methanol,ethanol,chloroform,,toluene,acetone,heptane,hexane,cyclohexane,,xylene and ether
Viscosity 1.23 mPa.s (20°C)
Formula Weight 108.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl chloroacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Toxic if swallowed.
May cause respiratory irritation.
Causes serious eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Fatal in contact with skin.
May cause
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi
EINECSNumber 202-501-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.