Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Methyl Cis-9,10-Methylene-Octadecanoate, Unsaturated Fatty Acid, 25 mg

Catalog No. 502706037
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6037 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6037 Supplier Abcam Supplier No. AB14407625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard for bacterial cyclopropane fatty acid synthesis assays with >98% purity.

  • Molecular Weight: 310.5 Da
  • Purity: >98%
  • Applications: Standard for bacterial cyclopropane fatty acid synthesis assays
  • Composition: Unsaturated fatty acid; product of cyclopropane synthase catalyzed S-adenosylmethionine methylation to the cis double bond of oleic acid
  • Constituents: 100% Methyl cis-9,10-methylene-octadecanoate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Methyl cis-9,10-methylene-octadecanoate
Content And Storage Store at -20°C.
Substrate Properties Lipid-based Substrate
Buffer 100% Methyl cis-9,10-methylene-octadecanoate
Formulation 100% Methyl cis-9,10-methylene-octadecanoate
Quantity 25 mg
For Use With (Application) bacterial cyclopropane fatty acid synthesis assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.