Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Coumalate 96.0+%, TCI America™
SDP

Catalog No. C173425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C173425G 25 g
C17345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C173425G Supplier TCI America Supplier No. C173425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6018-41-3
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00010120
InChI Key HHWWWZQYHPFCBY-UHFFFAOYSA-N
Synonym methyl coumalate, methyl 2-oxo-2h-pyran-5-carboxylate, coumalic acid, methyl ester, 5-carbomethoxy-2-pyrone, 2h-pyran-5-carboxylic acid, 2-oxo-, methyl ester, coumalic acid methyl ester, unii-1s7tmf0r8t, methyl 2-pyrone-5-carboxylate, methyl 2-oxopyran-5-carboxylate, 1s7tmf0r8t
PubChem CID 80113
IUPAC Name methyl 6-oxopyran-3-carboxylate
SMILES COC(=O)C1=COC(=O)C=C1

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 260°C
Quantity 25 g
Formula Weight 154.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Coumalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.