Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Crotonate 98.0+%, TCI America™
SDP

Catalog No. C0419100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0419100ML 100 mL
C041925ML 25 mL
C0419500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0419100ML Supplier TCI America Supplier No. C0419100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-43-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009287
InChI Key MCVVUJPXSBQTRZ-ONEGZZNKSA-N
Synonym methyl crotonate, e-methyl but-2-enoate, trans-methyl crotonate, methyl e-crotonate, methyl 2-butenoate, methyl trans-crotonate, 2-butenoic acid, methyl ester, 2e, trans-2-butenoic acid methyl ester, methyl trans-2-butenoate, e-crotonic acid methyl ester
PubChem CID 638132
IUPAC Name methyl (2E)-but-2-enoate
SMILES COC(=O)\C=C\C

Specifications

Color Colorless
Boiling Point 120°C
Quantity 100 mL
UN Number 3272
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Crotonate
EINECSNumber (2)-0966
RTECSNumber GQ5710000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.