Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Cyclopropanecarboxylate 97.0+%, TCI America™
SDP

Catalog No. C1607250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1607250ML 250 mL
C160725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1607250ML Supplier TCI America Supplier No. C1607250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2868-37-3
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00001280
InChI Key PKAHQJNJPDVTDP-UHFFFAOYSA-N
Synonym methyl cyclopropane carboxylate, cyclopropanecarboxylic acid, methyl ester, methylcyclopropanecarboxylate, cyclopropanecarboxylic acid methyl ester, methyl cyclopropylcarboxylate, unii-5uq53f585u, mcpc, pubchem11835, ksc490a7l, 1-methylcyclopropanecarboxylate
PubChem CID 76122
IUPAC Name methyl cyclopropanecarboxylate
SMILES COC(=O)C1CC1

Specifications

Color Yellow
Boiling Point 117°C
Quantity 250 mL
UN Number 1992
Formula Weight 100.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Cyclopropanecarboxylate
RTECSNumber GZ2017500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.