Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl β-D-thiogalactoside, 98%, Thermo Scientific™

Catalog No. AC459450010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459450010 1 g
AC459452500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459450010 Supplier Thermo Scientific Chemicals Supplier No. 459450010
Add to Cart

Chemical Identifiers

CAS 155-30-6
Molecular Formula C7H14O5S
Molecular Weight (g/mol) 210.25
InChI Key LZFNFLTVAMOOPJ-PZRMXXKTSA-N
Synonym methyl-beta-d-thiogalactoside, methyl-1-thio-beta-d-galactopyranoside, methyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol, b-d-thiomethylgalactoside, methyl b-d-thiogalactoside, galactose-derived compound 19, methyl beta-d-thio-galactopyranoside, methyl 1-thio-??-d-galactopyranoside, 1-s-methyl-1-thio-beta-d-galactopyranose
PubChem CID 10036019
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol
SMILES CSC1C(C(C(C(O1)CO)O)O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 210.25
Percent Purity 98%
Chemical Name or Material Methyl β-D-thiogalactoside
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Causes damage to organs.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 205-842-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.