Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Dacthal, SPEX CertiPrep™
SDP

Catalog No. S1065 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
S-1065 1 mL
1 options

Catalog No. S-1065

Supplier: SPEX CertiPrep S1065

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Methyl dacthal1861-32-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 1861-32-1 , 67-56-1
Molecular Formula C10H6Cl4O4
Molecular Weight (g/mol) 331.95
MDL Number MFCD00014902
InChI Key NPOJQCVWMSKXDN-UHFFFAOYSA-N
PubChem CID 2943
ChEBI CHEBI:34664
IUPAC Name 1,4-dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Methods For US EPA Method 8081
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methanol
Quantity 1 mL
Chemical Name or Material Methyl dacthal
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.