Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Difluoro(fluorosulfonyl)acetate 97.0+%, TCI America™
SDP

Catalog No. F031125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F031125G 25 g
F03115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F031125G Supplier TCI America Supplier No. F031125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 680-15-9
Molecular Formula C3H3F3O4S
Molecular Weight (g/mol) 192.108
MDL Number MFCD00144316
InChI Key GQJCAQADCPTHKN-UHFFFAOYSA-N
Synonym methyl 2,2-difluoro-2-fluorosulfonyl acetate, methylfluorosulphonyldifluoroacetate, methyl difluoro fluorosulfonyl acetate, methyl fluorosulfonyldifluoroacetate, methyl fluorosulphonyl difluoro acetate, methyl fluorosulphonyldifluoroacetate, methyl 2-fluorosulfonyl difluoroacetate, fluorosulfonyldifluoroacetic acid methyl ester, 2,2-difluoro-2-fluorosulfonylacetic acid methyl ester, difluoro fluorosulfonyl acetic acid methyl ester
PubChem CID 547667
IUPAC Name methyl 2,2-difluoro-2-fluorosulfonylacetate
SMILES COC(=O)C(F)(F)S(=O)(=O)F

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 g
UN Number 2920
Formula Weight 192.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Difluoro(fluorosulfonyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.