Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl diphenylphosphite, Thermo Scientific™

Catalog No. p-7049961
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3209703 1 g
AA3209706 5 g
AA3209714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3209703 Supplier Thermo Scientific Chemicals Supplier No. 3209703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3577-87-5
Molecular Formula C13H13O3P
Molecular Weight (g/mol) 248.22
MDL Number MFCD00014886
InChI Key NYCZNDFWFCCTPA-UHFFFAOYSA-N
Synonym diphenyl methyl phosphite, methyldiphenylphosphite, methoxydiphenoxyphosphine, methoxy-diphenoxy-phosphane, methyl diphenylphosphite 1g, phosphorous acid, methyl diphenyl ester
PubChem CID 520764
IUPAC Name methyl diphenyl phosphite
SMILES COP(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Quantity 1 g
UN Number UN3278
Sensitivity Hygroscopic
Formula Weight 248.21
Physical Form Liquid
Chemical Name or Material Methyl diphenylphosphite
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.