Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Gallate 98.0+%, TCI America™
SDP

Catalog No. G0017100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0017100G 100 g
G001725G 25 g
G0017500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0017100G Supplier TCI America Supplier No. G0017100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99-24-1
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.147
MDL Number MFCD00002194
InChI Key FBSFWRHWHYMIOG-UHFFFAOYSA-N
Synonym methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
PubChem CID 7428
IUPAC Name methyl 3,4,5-trihydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 100 g
Formula Weight 184.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Gallate
EINECSNumber (3)-1583
RTECSNumber LW8000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.