Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Methyl Heneicosanoate, >99% Purity, 340.6 Da, 100 mg

Catalog No. 502706065
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6065 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6065 Supplier Abcam Supplier No. AB143910100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Very long chain fatty acid methyl ester with >99% purity.

  • Molecular Weight: 340.6 Da
  • Purity: >99%
  • Composition: 100% Methyl heneicosanoate
  • Storage: Store at Ambient
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Buffer 100% Methyl heneicosanoate
Shipping Condition Ambient
Substrate Properties Lipid-based Substrate
Quantity 100 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.