Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Hesperidine 90.0+%, TCI America™
SDP

Catalog No. M033825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M033825G 25 g
M03385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M033825G Supplier TCI America Supplier No. M033825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 11013-97-1
Molecular Formula C29H36O15
MDL Number MFCD01741310
Synonym Vitamin P (Water soluble)

Specifications

Quantity 25 g
Formula Weight 624.59
Percent Purity ≥90.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Hesperidine
EINECSNumber (9)-1458
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.