Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Indole-3-acetate 98.0+%, TCI America™
SDP

Catalog No. M260525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M260525G 25 g
M26055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M260525G Supplier TCI America Supplier No. M260525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1912-33-0
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD00022749
InChI Key KTHADMDGDNYQRX-UHFFFAOYSA-N
Synonym Indole-3-acetic Acid Methyl Ester, IAA Methyl Ester
PubChem CID 74706
ChEBI CHEBI:72782
IUPAC Name methyl 2-(1H-indol-3-yl)acetate
SMILES COC(=O)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 51°C
Boiling Point 160°C
Quantity 25 g
Formula Weight 189.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Indole-3-acetate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.