Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Iodide, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18611701 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
60 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-701 25 g Amber Glass Bottle
18-611-700 100 g Amber Glass Bottle
18-611-702 500 g Amber Glass Bottle
18-611-703 60 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-701 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M127025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74-88-4
Molecular Formula CH3I
Molecular Weight (g/mol) 141.94
InChI Key INQOMBQAUSQDDS-UHFFFAOYSA-N
IUPAC Name iodomethane
SMILES CI

Specifications

CAS Max % 1
Boiling Point 43°C/109.4°F
Packaging Amber Glass Bottle
Quantity 25 g
Residue after Evaporation 0.0001
Percent Purity 0.995
Grade Reagent
Physical Form Liquid
Chemical Name or Material Methyl Iodide
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.