Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Isonicotinate 99.0+%, TCI America™
SDP

Catalog No. I0141100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0141100G 100 g
I014125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0141100G Supplier TCI America Supplier No. I0141100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9-8-2459
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006427
InChI Key OLXYLDUSSBULGU-UHFFFAOYSA-N
Synonym methyl isonicotinate, isonicotinic acid methyl ester, 4-pyridinecarboxylic acid, methyl ester, 4-picolinic acid methyl ester, 4-methoxycarbonylpyridine, methylisonicotinate, methyl 4-pyridinecarboxylate, 4-carbomethoxypyridine, isonicotinic acid, methyl ester, i-nicotinic acid, methyl ester
PubChem CID 227085
IUPAC Name methyl pyridine-4-carboxylate
SMILES COC(=O)C1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 209°C
Quantity 100 g
Formula Weight 137.14
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Methyl Isonicotinate
EINECSNumber (5)-0733
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.