Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl isovalerate, 98%

Catalog No. AAL0624130
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0624130 250 g
AAL0624118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0624130 Supplier Thermo Scientific Chemicals Supplier No. L0624130
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 556-24-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-24-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00042866
InChI Key OQAGVSWESNCJJT-UHFFFAOYSA-N
Synonym methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
PubChem CID 11160
IUPAC Name methyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC

Specifications

Density 0.88
Boiling Point 115°C to 117°C
Flash Point 16°C (60°F)
Packaging Glass bottle
Refractive Index 1.3936
Quantity 250 g
UN Number UN2400
Beilstein 1699922
Specific Gravity 0.88
Formula Weight 116.17
Percent Purity 98%
Chemical Name or Material Methyl isovalerate
Show More Show Less
Hazard Category H225
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 209-117-3
RTECSNumber NY1510000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.