Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl isovalerate, 98%, Thermo Scientific™

Catalog No. AAA12819AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12819AE 100 mL
AAA12819AP 500 mL
AAA128190F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12819AE Supplier Thermo Scientific Chemicals Supplier No. A12819AE

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-24-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00042866
InChI Key OQAGVSWESNCJJT-UHFFFAOYSA-N
Synonym methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
PubChem CID 11160
IUPAC Name methyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC

Specifications

Boiling Point 115°C to 117°C
Flash Point 16°C (61°F)
Quantity 100 mL
UN Number UN2400
Formula Weight 116.1
Percent Purity 98%
Chemical Name or Material Methyl isovalerate
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ISOVALERATE
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.