Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl malonyl chloride, 97%

Catalog No. p-7049742
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2082306 5 g
AA2082314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2082306 Supplier Thermo Scientific Chemicals Supplier No. 02082306
Only null left
Add to Cart

CAS: 37517-81-0 | C4H5ClO3 | 136.531 g/mol

Methyl malonyl chloride is used as pharmaceutical intermediate. Used in anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37517-81-0
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00013657
InChI Key UTBCRHAMJFMIIR-UHFFFAOYSA-N
PubChem CID 123460
IUPAC Name methyl 3-chloro-3-oxopropanoate
SMILES COC(=O)CC(=O)Cl

Specifications

Color Colorless
Density 1.27 g/mL
Boiling Point 58°C (12 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.4325
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate
Solubility Information Reacts with water.
Formula Weight 136.53
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl malonyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 253-540-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.