Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange Indicator, Reag. Ph. Eur., Solstice

Catalog No. 60046136 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-136 100 g Glass Bottle
60-046-137 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-136 Supplier Solstice Supplier No. 32624100G

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym Helianthin
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

UN Number UN3143
Beilstein 4732884
Formula Weight 327.33g/mol
Packaging Glass Bottle
Suitability Complies for sensitivity
Visual Transition Interval 3.0 to 4.4
Color Orange
pH 3.0 to 4.4 (pink to yellow)
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Methyl Orange, Reag. Ph. Eur.
Show More Show Less
Hazard Category Danger
Hazard Statement P301 + P310
Precautionary Statement H301
DOTInformation 6.1/PGIII
EINECSNumber 208-925-3
RTECSNumber DB6327000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.