Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Propyl Disulfide 97.0+%, TCI America™
SDP

Catalog No. M17815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17815G 5 g
M178125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17815G Supplier TCI America Supplier No. M17815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2179-60-4
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.244
MDL Number MFCD00013400
InChI Key PUCHCUYBORIUSM-UHFFFAOYSA-N
Synonym methyl propyl disulfide, disulfide, methyl propyl, 1-methyldisulfanyl propane, 2,3-dithiahexane, methyl propyl disulphide, methyldithiopropane, propyl methyl disulfide, methyl n-propyl disulfide, unii-8m12k3fu0t, methylpropyldisulfide
PubChem CID 16592
IUPAC Name 1-(methyldisulfanyl)propane
SMILES CCCSSC

Specifications

Color Yellow
Boiling Point 71°C
Quantity 5 g
UN Number 1993
Formula Weight 122.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Propyl Disulfide
EINECSNumber (2)-0477
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.