Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylate 98.0+%, TCI America™
SDP

Catalog No. D256225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D256225G 25 g
D25625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D256225G Supplier TCI America Supplier No. D256225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52373-72-5
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00066527
InChI Key DOWWCCDWPKGNGX-RXMQYKEDSA-N
Synonym (+)-2,3-O-Isopropylidene-D-glyceric Acid Methyl Ester, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, (+)-Methyl 2,3-O-Isopropylidene-D-glycerate
PubChem CID 853166
IUPAC Name methyl (4R)-2,2-dimethyl-1,3-dioxolane-4-carboxylate
SMILES CC1(OCC(O1)C(=O)OC)C

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.