Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red Indicator, 0.1% w/v in 60% Methanol, Reagents

Catalog No. 01336195
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-195 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-195 Supplier Reagents Holdings Llc Supplier No. C83021NE1C
Add to Cart
Add to Cart

Methyl Red Indicator, 0.1% w/v in 60% Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methyl Red Indicator, 0.1% w/v in 60% Methanol
Boiling Point 71°C
CAS 67-56-1, 7732-18-5, 7732-18-5
Synonym Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid
Percent Purity 0.10%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.91
Color Red
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.