Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(+)-3-Hydroxybutyrate 98.0+%, TCI America™
SDP

Catalog No. H070425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H070425ML 25 mL
H07045ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H070425ML Supplier TCI America Supplier No. H070425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53562-86-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00064461
InChI Key LDLDJEAVRNAEBW-UHFFFAOYNA-N
Synonym methyl s-+-3-hydroxybutyrate, s-methyl 3-hydroxybutanoate, methyl s-3-hydroxybutyrate, s-3-hydroxybutanoic acid methyl ester, unii-g10281a5ah, methyl 3s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, methyl ester, 3s, methyl-s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid methyl ester, s-3-hydroxybutyric acid methyl ester
PubChem CID 6950307
IUPAC Name methyl 3-hydroxybutanoate
SMILES COC(=O)CC(C)O

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 mL
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(+)-3-Hydroxybutyrate
EINECSNumber (2)-1430
RTECSNumber ET4700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.