Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Sulfone, Crystal, Spectrum™ Chemical
SDP

Catalog No. 18621091 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
12 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-091 500 g
18-621-090 100 g
18-621-092 5 kg
18-621-093 12 kg
18-621-094 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-621-091 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M2125500GM
Add to Cart
Add to Cart

Synonym: Dimethyl Sulfone, Methylsulfonylmethane, Dimethylsulfone, Sulphonylbismethane

Chemical Identifiers

CAS 67-71-0

Specifications

Packaging Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.