Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide-d6, for NMR, contains 1 v/v% TMS, 99.9% atom D, ACROS Organics™

Click to view available options
Quantity:
10mL
25mL
50mL
Packaging:
Glass bottle
2206-27-1
C2H6OS
84.166
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide
75151
trideuterio(trideuteriomethylsulfinyl)methane
CS(=O)C
2206-27-1
C2H6OS
84.166
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide
75151
trideuterio(trideuteriomethylsulfinyl)methane
CS(=O)C
Color Colorless
Viscosity 2.4 mPa.s (20°C),2.4 mPa.s (20°C)
Melting Point 20.2°C
Flash Point 88°C
Specific Gravity 1.19
Boiling Point 190.0°C
Quantity 10mL
Packaging Glass bottle
Deuterium (D) 99.85% min.
Water 0.03% max. (Coulometric)
Density 1.1900g/mL
CAS Min % 99.85
CAS Max % 100.0
Linear Formula (CD3)2SO
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Formula Weight 84.18
Physical Form Liquid
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6, 99.9 atom % D
Show More Show Less

HYGROSCOPIC

EINECSNumber : 218-617-0

TSCA : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.