Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide-d6, for NMR, packaged in 1.00 ml ampoules, 99.9 atom % D

Catalog No. AC217330100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC217330100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC217330100 Supplier Thermo Scientific Chemicals Supplier No. 217330100
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Packaging Ampule
Quantity 10 mL
Flash Point 88°C
Specific Gravity 1.19
Viscosity 2.4 mPa.s (20°C)
Deuterium (D) 99.85% min.
Density 1.1900g/mL
Water 0.03% max. (Coulometric)
Melting Point 20.2°C
Boiling Point 190.0°C
Linear Formula (CD3)2SO
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Formula Weight 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6, 99.9 atom % D
Show More Show Less
Hazard Category HYGROSCOPIC

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.